Skip to Content
Merck
Potassium dichromate solution c(K2Cr2O7) = 1/24 mol/l (0.25 N), Titripur®

Potassium dichromate solution

Potassium dichromate solution c(K2Cr2O7) = 1/24 mol/l (0.25 N), Titripur®
Synonym(s):
Chromic acid solution

Products

Product Category
Brand
Physical Form
Product Line
Quality Segment
Special Grade
Technique