Skip to Content
Merck
Splitomicin ≥98% (HPLC), powder

Splitomicin

Splitomicin ≥98% (HPLC), powder
Synonym(s):
1,2-Dihydro-3H-naphtho[2,1-b]pyran-3-one
Empirical Formula (Hill Notation):
C13H10O2
CAS Number:
Molecular Weight:
198.22

Products

Product Category
Brand
Color
Physical Form
Purity
Quality Segment