Skip to Content
Products
Cart
0
TH
EN
Products
Login / Register
Order Lookup
Quick Order
Cart
0
Home
Search Results
Lipases: from production to applications
Lipases: from production to applications
Separation & Purification Reviews (2020)
Melani N, et al.
An unknown error has occured.
ไทย - TH
English - EN
Learn More