Merck
  • PIBF+ extracellular vesicles from mouse embryos affect IL-10 production by CD8+ cells.
PIBF+ extracellular vesicles from mouse embryos affect IL-10 production by CD8+ cells.
Scientific reports (2018-03-20)
Eva Pallinger, Zoltan Bognar, Agnes Bogdan, Timea Csabai, Hajnalka Abraham, Julia Szekeres-Bartho
ABSTRACT
Earlier evidence suggests, that the embryo signals to the maternal immune system. Extracellular vesicles (EVs) are produced by all types of cells, and because they transport different kinds of molecules from one cell to the other, they can be considered as means of intercellular communication. The aim of this work was to test, whether the embryo is able to produce sufficient amounts of EVs to alter the function of peripheral lymphocytes. Embryo-derived EVs were identified by their Annexin V biding capacity, and sensitivity to Triton X dependent lysis, using flow cytometry. Transmission electron microscopy was used to detect EVs at the implantation site. Progesterone-induced blocking factor (PIBF) expression in embryo-derived EVs was demonstrated with immuno-electron microscopy. The % of IL-10 + murine lymphocytes was determined by flow cytometry. EVs were present in embryo culture media, but not in empty media. Mouse embryo-derived EVs adhere to the surface of both CD4+ and CD8+ murine peripheral T lymphocytes, partly, via phosphatidylserine binding. The number of IL-10+ murine peripheral CD8+ cells increases in the presence of embryo-derived EVS, and this effect is counteracted by pre-treatment of EVs with an anti-PIBF antibody, suggesting that the embryo communicates with the maternal immune system via EVs.
Sigma-Aldrich
Anti-Phosphatidylserine Receptor antibody produced in rabbit, 1-1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.