Merck
  • 3,3'-Bicarbazole-Based Host Molecules for Solution-Processed Phosphorescent OLEDs.
3,3'-Bicarbazole-Based Host Molecules for Solution-Processed Phosphorescent OLEDs.
Molecules (Basel, Switzerland) (2018-04-13)
Jungwoon Kim, Suhan Lee, Jaemin Lee, Eunhee Lim, Byung Jun Jung
ABSTRACT
Solution-processed organic light-emitting diodes (OLEDs) are attractive due to their low-cost, large area displays, and lighting features. Small molecules as well as polymers can be used as host materials within the solution-processed emitting layer. Herein, we report two 3,3'-bicarbazole-based host small molecules, which possess a structural isomer relationship. 9,9'-Di-4-n-butylphenyl-9H,9'H-3,3'-bicarbazole (BCz-nBuPh) and 9,9'-di-4-t-butylphenyl-9H,9'H-3,3'-bicarbazole (BCz-tBuPh) exhibited similar optical properties within solutions but different photoluminescence within films. A solution-processed green phosphorescent OLED with the BCz-tBuPh host exhibited a high maximum current efficiency and power efficiency of 43.1 cd/A and 40.0 lm/W, respectively, compared to the device with the BCz-nBuPh host.
Sigma-Aldrich
4-tert-Butyliodobenzene, 98%
Pricing and availability is not currently available.