Merck
  • High glucose concentration suppresses a SIRT2 regulated pathway that enhances neurite outgrowth in cultured adult sensory neurons.
High glucose concentration suppresses a SIRT2 regulated pathway that enhances neurite outgrowth in cultured adult sensory neurons.
Experimental neurology (2018-08-14)
Emily Schartner, Mohammad Golam Sabbir, Ali Saleh, Rafaela Vieira Silva, Subir Roy Chowdhury, Darrell R Smith, Paul Fernyhough
ABSTRACT
In peripheral nerve under hyperglycemic conditions high flux of d-glucose through the polyol pathway drives an aberrant redox state contributing to neurodegeneration in diabetic sensorimotor polyneuropathy (DSPN). Sirtuins, including SIRT2, detect the redox state via the NAD+/NADH ratio to regulate mitochondrial function via, in part, AMP-activated protein kinase (AMPK) and peroxisome proliferator-activated receptor γ coactivator 1-α (PGC-1α). In adult dorsal root ganglia (DRG) sensory neurons mitochondrial dysfunction has been proposed as an etiological factor in dying-back neuropathy in diabetes. We tested the hypothesis that a high concentration of d-glucose depleted SIRT2 expression via enhancement of polyol pathway activity. We posited that this would lead to impaired mitochondrial function and suppression of neurite outgrowth in cultured sensory neurons. The use of dominant negative mutants or neurons from SIRT2 knockout (KO) mice to block SIRT2 signaling revealed that neurons derived from control or type 1 diabetic rodents required SIRT2 for optimal neurite outgrowth. Over-expression of WT-SIRT2 elevated neurite outgrowth in normal and diabetic cultures. SIRT2 protein isoforms 2.1 and 2.2 were reduced by 20-30% in DRG of type 1 diabetic mice (p < .05). After 72 h exposure to high d-glucose (25 mM vs 5 mM) cultured sensory neurons showed a significant 2-fold (p < .05) decrease in SIRT2 expression, P-AMPK, levels of respiratory Complexes II/III and respiratory capacity. DRG neurons expressed aldose reductase and the aforementioned deficits were prevented by treatment with aldose reductase inhibitors (lidorestat or sorbinil) or sorbitol dehydrogenase inhibitor (SDI-158). In cultures derived from type 1 diabetic rats treatment with SDI-158 elevated expression of SIRT2, P-AMPK/PGC-1α and neurite outgrowth (p < .05). SIRT2 KO neurons exhibited deficits in the LKB-1/AMPK/PGC-1α pathway and mitochondrial function. In cultured neurons the SIRT2 pathway enhances axonal outgrowth and this signaling axis encompassing activation of AMPK/PGC-1α is impaired in DSPN, in part, due to enhanced polyol pathway activity caused by hyperglycemia.
Sigma-Aldrich
DL-Glyceraldehyde, ≥90% (GC)
Sigma-Aldrich
SIGMAFAST DAB with Metal Enhancer, tablet
Sigma-Aldrich
Sorbinil, ≥98% (HPLC)