Merck
  • Mir-758-5p Suppresses Glioblastoma Proliferation, Migration and Invasion by Targeting ZBTB20.
Mir-758-5p Suppresses Glioblastoma Proliferation, Migration and Invasion by Targeting ZBTB20.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2018-08-14)
Ji Liu, Jian Jiang, Xiaobo Hui, Weijie Wang, Dazhao Fang, Lianshu Ding
ABSTRACT
To determine the cellular functions and clinical significance of micro-758-5p (miR-758-5p) in glioblastoma (GBM) by targeting zinc finger and BTB domain-containing protein 20 (ZBTB20). Fifty-five paired GBM tissues and adjacent normal tissues, GBM cell lines (U118, LN-299, H4, A172, U87-MG, and U251), and normal human astrocyte cell line (HEB) were used. miR-758-5p mimics, ZBTB20 siRNA, and pcDNA3.1-ZBTB20 were transiently transduced into cancer cells independently or together. qRT-PCR was conducted to analyze the expression of miR-758-5p and ZBTB20. Luciferase reporter assays were performed to determine the effect of miR-758-5p on ZBTB20. Western blot was applied to measure the expression of ZBTB20, PCNA, and cleaved caspase3. Cell Counting Kit-8 (CCK8), colony formation, FACS, and Transwell assays were carried out to detect cellular proliferation, apoptosis, migration, and invasion. Xenograft experiments were implemented to evaluate tumor growth and metastasis in vivo. miR-758-5p was significantly downregulated in GBM tissues and cell lines compared with that in adjacent normal tissues and HEB cells. miR-758-5p overexpression inhibited the proliferation, migration, and invasion of GBM cells and induced apoptosis by regulating the ZBTB20 expression. Pearson correlation analysis also confirmed that miR-758-5p was inversely correlated with ZBTB20 in GBM tissues. miR-758-5p suppressed tumor growth and metastasis in vivo. The restored ZBTB20 expression partially rescued the miR-758-5p-induced inhibition of GBM cell proliferation, migration, and invasion. Kaplan-Meier curve analysis revealed that a high miR-758-5p expression indicated an enhanced prognosis of patients with GBM. miR-758-5p suppressed GBM progression by targeting ZBTB20. The miR-758-5p/ZBTB20 axis might be a promising therapeutic target for GBM treatment.
Sigma-Aldrich
MISSION® esiRNA, targeting human ZBTB20