Merck
  • Calcification in coronary artery disease can be reversed by EDTA-tetracycline long-term chemotherapy
Calcification in coronary artery disease can be reversed by EDTA-tetracycline long-term chemotherapy
Pathophysiology : The Official Journal of the International Society for Pathophysiology / ISP (2004)
Maniscalco BS and Taylor KA