Merck
  • An overview of the effect of linoleic and conjugated-linoleic acids on the growth of several human tumor cell lines
An overview of the effect of linoleic and conjugated-linoleic acids on the growth of several human tumor cell lines
International Journal of Cancer. Journal International Du Cancer (2004)
Maggiora M, et al.