Merck
  • An ex vivo method for evaluating the biocompatibility of neural electrodes in rat brain slice cultures
An ex vivo method for evaluating the biocompatibility of neural electrodes in rat brain slice cultures
Journal of Neuroscience Methods (2004)
Koeneman BA, et al.