Merck
  • Signal transduction for proteinase-activated receptor-2-triggered prostaglandin E2 formation in human lung epithelial cells
Signal transduction for proteinase-activated receptor-2-triggered prostaglandin E2 formation in human lung epithelial cells
Journal of Pharmacology and Experimental Therapeutics (2005)
Kawao N, et al.