- Coexpression of keratin-and vimentin-type intermediate filaments in human metastatic carcinoma cells
Coexpression of keratin-and vimentin-type intermediate filaments in human metastatic carcinoma cells
Proceedings of the National Academy of Sciences of the USA (1983)
Ramaekers FC, et al.