Merck
  • A Stretchable and Self-Healing Energy Storage Device Based on Mechanically and Electrically Restorative Liquid-Metal Particles and Carboxylated Polyurethane Composites.
A Stretchable and Self-Healing Energy Storage Device Based on Mechanically and Electrically Restorative Liquid-Metal Particles and Carboxylated Polyurethane Composites.
Advanced materials (Deerfield Beach, Fla.) (2018-11-06)
Sangbaek Park, Gurunathan Thangavel, Kaushik Parida, Shaohui Li, Pooi See Lee
ABSTRACT
Stretchable and self-healing (SH) energy storage devices are indispensable elements in energy-autonomous electronic skin. However, the current collectors are not self-healable nor intrinsically stretchable, they mostly rely on strain-accommodating structures that require complex processing, are often limited in stretchability, and suffer from low device packing density and fragility. Here, an SH conductor comprising nickel flakes, eutectic gallium indium particles (EGaInPs), and carboxylated polyurethane (CPU) is presented. An energy storage device is constructed by the two SH electrodes assembled with graphene nanoplatelets sandwiching an ionic-liquid electrolyte. An excellent electrochemical healability (94% capacity retention upon restretching at 100% after healing from bifurcation) is unveiled, stemming from the complexation modulated redox behavior of EGaIn in the presence of the ligand bis(trifluoromethanesulfonyl)imide, which enhances the reversible Faradaic reaction of Ga. Self-healing can be achieved where the damaged regions are electrically restored by the flow of liquid metal and mechanically healing activated by the interfacial hydrogen bonding of CPU with an efficiency of 97.5% can be achieved. The SH conductor has an initial conductivity of 2479 S cm-1 that attains a high stretchability with 700% strain, it restores 100% stretchability even after breaking/healing with the electrical healing efficiency of 75%.