Merck
  • Nylon surface modification. Part 1. Targeting the amide groups for selective introduction of reactive functionalities
Nylon surface modification. Part 1. Targeting the amide groups for selective introduction of reactive functionalities
Polymer (2006)
Jia X, et al.