Merck
  • Behavioral and Biochemical Features of the Course and Surgical Treatment of Experimental Obstructive Hydrocephalus in Young Rats.
Behavioral and Biochemical Features of the Course and Surgical Treatment of Experimental Obstructive Hydrocephalus in Young Rats.
Developmental neuroscience (2019-04-19)
Marcelo Volpon Santos, Luiza da Silva Lopes, Hélio Rubens Machado, Ricardo Santos de Oliveira
ABSTRACT
Hydrocephalus is a multifactorial disease, affecting the dynamics of cerebrospinal fluid (CSF) and leading to severe neurological impairment in children; in spite of the recent advances in hydrocephalus research, it has many physiopathological aspects that still remain poorly understood, especially after treatment. To analyze the clinical, radiological, histopathological, and biochemical aspects of kaolin-induced hydrocephalus in an experimental model, both in the acute phase and after shunt treatment, by means of behavioral tests, magnetic resonance imaging (MRI) scans, histopathological studies, and level of inflammatory interleukins in the CSF. Seven-day-old Wistar rats were used and subdivided into three subgroups: treated hydrocephalic (n = 24), untreated hydrocephalic (n = 17), and controls (n = 5). The hydrocephalic groups underwent cisternal injection of 15% kaolin for induction of hydrocephalus at 7 days of age. The treated group was submitted to a ventricular-subcutaneous shunt (VSCS) 1 week after induction. All animals were euthanized at 21 days of age. They underwent motor function and memory testing as well as brain MRI scans. Histopathological analysis for glial fibrillary acidic protein and Ki-67 was done, and CSF was collected for measurement of IL-1β, IL-6, and TNF-α. The average time to reach the water maze platform was highest in the untreated hydrocephalic group. The magnetization transfer rates were 37.21 and 33.76 before and after shunting, respectively. The mean astrocyte counts were 2.45, 1.36, and 90.5 for shunted, untreated, and control animals, respectively. The mean CSF IL-1β concentrations were 62.3 and 249.6 pg/mL, the average IL-6 levels were 104.2 and 364.7 pg/mL, and the average TNF-α values were 4.9 and 170.5 pg/mL for the treated hydrocephalic group and the untreated group, respectively. Pups treated with a CSF shunt showed better performance on memory tests. VSCS did not revert demyelination caused by hydrocephalus. Likewise, reactive astrocytosis and cell proliferation over the germinal matrix were not reversed after shunting. Hydrocephalic animals had raised levels of inflammatory interleukins, which returned to normal after treatment.
Product Description
Supelco
4-Bromofluorobenzene, analytical standard
Pricing and availability is not currently available.