Merck
  • Extended delivery of non-steroidal anti-inflammatory drugs through contact lenses loaded with Vitamin E and cationic surfactants.
Extended delivery of non-steroidal anti-inflammatory drugs through contact lenses loaded with Vitamin E and cationic surfactants.
Contact lens & anterior eye : the journal of the British Contact Lens Association (2019-05-16)
Cesar Torres-Luna, Naiping Hu, Tejaswi Tammareddy, Roman Domszy, Jeffrey Yang, Nam Sun Wang, Arthur Yang
ABSTRACT
The purpose of this study is to extend drug release from ACUVUE Oasys® and ACUVUE TruEye® silicone hydrogel contact lenses by incorporation of vitamin E in conjunction with a cationic surfactant. In ACUVUE Oasys® and ACUVUE TruEye®, the release of ketorolac tromethamine and flurbiprofen sodium is extended from hours to several days for 11% and 21% vitamin E, (weight of vitamin E / weight of dry lens) but with a considerable reduction in the amount of drug released. Cetalkonium chloride and stearylamine increased the drug loading capacity which was otherwise compromised by the addition of vitamin E in the contact lenses. In the case of diclofenac sodium, a sustained release over 150 h for both contact lenses can be achieved. It was found that the release-time-increase factor due to vitamin E has a linear dependence with the octanol-water partition coefficient of the drug in ACUVUE Oasys®. The results in this study show that contact lenses loaded with vitamin E in conjunction with cationic surfactants achieved sustained release of non-steroidal anti-inflammatory drugs (NSAIDs) within the therapeutic window.
Sigma-Aldrich
Octadecylamine, ≥99% (GC)
Pricing and availability is not currently available.