Merck
Disruption of Telomerase RNA Maturation Kinetics Precipitates Disease.
Molecular cell (2019-04-02)
Caitlin M Roake, Lu Chen, Ananya L Chakravarthy, James E Ferrell, Grazia D Raffa, Steven E Artandi
ABSTRACT
Mutations in RNA-processing enzymes are increasingly linked to human disease. Telomerase RNA and related noncoding RNAs require 3' end-processing steps, including oligoadenylation. Germline mutations in poly(A)ribonuclease (PARN) cause accumulation of extended human telomerase RNA (hTR) species and precipitate dyskeratosis congenita and pulmonary fibrosis. Here, we develop nascent RNAend-seq to measure processing rates of RNA precursors. We find that mature hTR derives from extended precursors but that in PARN-mutant cells hTR maturation kinetically stalls and unprocessed precursors are degraded. Loss of poly(A)polymerase PAPD5 in PARN-mutant cells accelerates hTR maturation and restores hTR processing, indicating that oligoadenylation and deadenylation set rates of hTR maturation. The H/ACA domain mediates hTR maturation by precisely defining the 3' end, recruiting poly(A)polymerase activity, and conferring sensitivity to PARN regulation. These data reveal a feedforward circuit in which post-transcriptional oligoadenylation controls RNA maturation kinetics. Similar alterations in RNA processing rates may contribute to mechanisms of RNA-based human disease.
Product Description
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
Anti-PAPD5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)