Merck
  • Novel anticholinesterases based on the molecular skeletons of furobenzofuran and methanobenzodioxepine.
Novel anticholinesterases based on the molecular skeletons of furobenzofuran and methanobenzodioxepine.
Journal of medicinal chemistry (2005-02-18)
Weiming Luo, Qian-sheng Yu, Ming Zhan, Damon Parrish, Jeffrey R Deschamps, Santosh S Kulkarni, Harold W Holloway, George M Alley, Debomoy K Lahiri, Arnold Brossi, Nigel H Greig
ABSTRACT
Reductive cyclization of 5-hydroxy-3-methyl-3-methoxycarbonylmethylenebenzofuran-2(3H)-one (4) gave 5-hydroxy-3a-methyl-2,3,3a,8a-tatrahydrofuro[2,3-b]benzofuran (5) and the rearrangement product 7-hydroxy-4,5-dihydro-2,5-methano-1,3-benzodioxepine (6). Reaction of compounds 5 and 6 with different isocyanates provided two series novel carbamates (7-12) whose structures were confirmed by X-ray crystallography. These were assessed for anticholinesterase action against freshly prepared human enzyme and proved to be potent inhibitors of either acetyl- (AChE) or butyrylcholinesterase (BChE) with specific compounds exhibiting remarkable selectivity. Because the two series of carbamates (7-12) differ in their phenolic moieties, their respective potency and selectivity for AChE versus BChE was governed by their N-substituted groups. This same characteristic was also present in a series of physovenine analogues (1, 13, 15, 17) and physostigmine analogues (2, 14, 16, 18). These structure-activity relations proved valuable in elucidating the mechanisms underpinning the interaction between carbamate-based cholinesterase inhibitors and their enzyme target. In addition, because physostigmine analogues have demonstrated activity in lowering the Alzheimer's disease protein, amyloid precursor protein (APP), examples of the two new series of carbamates were characterized in culture assays of quantifying cell viability and synthesis of APP.