Merck
The double-edged sword of activation-induced cytidine deaminase.
Journal of immunology (Baltimore, Md. : 1950) (2005-01-07)
Xiaosheng Wu, Pedro Geraldes, Jeffrey L Platt, Marilia Cascalho
ABSTRACT
Activation-induced cytidine deaminase (AID) is required for Ig class switch recombination, a process that introduces DNA double-strand breaks in B cells. We show in this study that AID associates with the DNA-dependent protein kinase catalytic subunit (DNA-PKcs) promoting cell survival, presumably by resolving DNA double-strand breaks. Wild-type cells expressing AID mutants that fail to associate with DNA-PKcs or cells deficient in DNA-PKcs or 53BP1 expressing wild-type AID accumulate gammaH2AX foci, indicative of heightened DNA damage response. Thus, AID has two independent functions. AID catalyzes cytidine deamination that originates DNA double-strand breaks needed for recombination, and it promotes DNA damage response and cell survival. Our results thus resolve the paradox of how B cells undergoing DNA cytidine deamination and recombination exhibit heightened survival and suggest a mechanism for hyperIgM type II syndrome associated with AID mutants deficient in DNA-PKcs binding.
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone 2-28-33, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-DNA-PK (Ab-2) Mouse mAb (18-2), liquid, clone 18-2, Calbiochem®
Pricing and availability is not currently available.