Merck
  • Neurofibromin regulates metabolic rate via neuronal mechanisms in Drosophila.
Neurofibromin regulates metabolic rate via neuronal mechanisms in Drosophila.
Nature communications (2021-07-15)
Valentina Botero, Bethany A Stanhope, Elizabeth B Brown, Eliza C Grenci, Tamara Boto, Scarlet J Park, Lanikea B King, Keith R Murphy, Kenneth J Colodner, James A Walker, Alex C Keene, William W Ja, Seth M Tomchik
ABSTRACT
Neurofibromatosis type 1 is a chronic multisystemic genetic disorder that results from loss of function in the neurofibromin protein. Neurofibromin may regulate metabolism, though the underlying mechanisms remain largely unknown. Here we show that neurofibromin regulates metabolic homeostasis in Drosophila via a discrete neuronal circuit. Loss of neurofibromin increases metabolic rate via a Ras GAP-related domain-dependent mechanism, increases feeding homeostatically, and alters lipid stores and turnover kinetics. The increase in metabolic rate is independent of locomotor activity, and maps to a sparse subset of neurons. Stimulating these neurons increases metabolic rate, linking their dynamic activity state to metabolism over short time scales. Our results indicate that neurofibromin regulates metabolic rate via neuronal mechanisms, suggest that cellular and systemic metabolic alterations may represent a pathophysiological mechanism in neurofibromatosis type 1, and provide a platform for investigating the cellular role of neurofibromin in metabolic homeostasis.
Sigma-Aldrich
Glycerol Standard Solution, 2.5 mg/ml equivalent triolein concentration
Sigma-Aldrich
Free Glycerol Reagent, used for quantitative enzymatic determination of glycerol
Sigma-Aldrich
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse, clone MAPK-YT, ascites fluid
Sigma-Aldrich
Triglyceride Reagent, used for quantitative enzymatic determination of triglyerides