Merck
  • Hypoxia-sensitive supramolecular nanogels for the cytosolic delivery of ribonuclease A as a breast cancer therapeutic.
Hypoxia-sensitive supramolecular nanogels for the cytosolic delivery of ribonuclease A as a breast cancer therapeutic.
Journal of controlled release : official journal of the Controlled Release Society (2020-01-20)
Xinghui Si, Sheng Ma, Yudi Xu, Dawei Zhang, Na Shen, Haiyang Yu, Yu Zhang, Wantong Song, Zhaohui Tang, Xuesi Chen
ABSTRACT
As the most common malignancy in women, breast cancer causes >40,000 deaths annually. Ribonuclease A (RNase), a new anti-cancer agent, has attracted intense interest due to its high efficacy and specificity. However, RNase suffers from instability, a short half-life in the circulation and poor membrane penetration. To overcome these challenges, we designed a supramolecular nanogel for the cytosolic delivery of RNase. The nanogels were fabricated using host-guest interactions between azobenzene (Azo) and β-cyclodextrin (βCD) conjugated to poly (L-glutamic acid)-graft-poly (ethylene glycol) methyl ether (PLG-g-mPEG). RNase could be loaded inside the nanogels in mild aqueous conditions. Following optimization, the RNase-loading content and efficiency of the nanogel were 23.5 wt% and 50.4%, respectively. In the presence of nitroreductase (NTR), the cross-linking point between Azo and βCD was destroyed due to the conformation transition of Azo, ensuring the hypoxia-sensitive release of cargo from the nanogels in tumors in which NTR is overexpressed. In vitro release profiles revealed that 75.0% of the RNase was released under hypoxic conditions in 72 h, whilst only 19.7% was released under normoxic conditions. Cytotoxicity assays showed that the RNase-loaded nanogels (nano-RNase) were more efficient in inhibiting the proliferation of 4T1 cells than free RNase. In vivo studies showed 68.7% tumor suppression rates (TSR %) in the nano-RNase treated group, whilst free RNase treatment led to a lack of tumor inhibition. To further enhance the hypoxia status of tumors, we combined nano-RNase with a nanoformulation of vascular disrupting agents PLG-g-mPEG/combretastatinA4 (nano-CA4) and obtained a TSR of 91.7%. The hypoxia-sensitive supramolecular nanogels provided a versatile platform for the delivery of RNase, highlighting its applicability for cancer therapy.