Merck
  • Capillary zone electrophoresis of oligosaccharides derivatized with N-(4-aminobenzoyl)-L-glutamic acid for ultraviolet absorbance detection.
Capillary zone electrophoresis of oligosaccharides derivatized with N-(4-aminobenzoyl)-L-glutamic acid for ultraviolet absorbance detection.
Journal of chromatography. A (1997-01-03)
J Plocek, M V Novotny
ABSTRACT
A charged and strongly UV-absorbing tag, N-(4-aminobenzoyl)-L-glutamic acid) (ABG), was coupled to oligosaccharides by reductive amination under mild conditions. The effectiveness of ABG as a derivatization agent is shown through the separation of isomaltooligosaccharides from a dextran hydrolysate. The minimum detectable quantities in the subpicomole range are demonstrated.
Sigma-Aldrich
N-(4-Aminobenzoyl)-L-glutamic acid, ≥98% (TLC)
Pricing and availability is not currently available.