Merck
  • Stimulation or Inhibition: Conflicting evidence for (+/-)-catechin's role as a chemical facilitator and disease protecting agent.
Stimulation or Inhibition: Conflicting evidence for (+/-)-catechin's role as a chemical facilitator and disease protecting agent.
Plant signaling & behavior (2009-12-22)
Harsh P Bais, L Venkatachalam, Meredith L Biedrzycki
ABSTRACT
The occurrence of plant hormesis is a poorly understood phenomenon, wherein low doses of phytotoxins unusually promote growth responses in higher plants. In contrast, negative plant-plant interactions mediated through secreted small molecular weight compounds initiate growth inhibitory responses. Studies related to (+/-)-catechin mediated allelopathy have transpired both novel information and generated significant controversy. Specifically, studies related to the phytotoxicity responses mediated by (+/-)-catechins have been seriously debated. The pronged opinion that (+/-)-catechin is phytotoxic versus non-phytotoxic relies more on the target plant systems and the conditions used to test phytotoxic responses. It is reported that lower than MIC dosage supplementation of (+/-)-catechin could promote growth responses in the model plant Arabidopsis thaliana. Furthermore, it was shown that sub-MIC levels of (+/-)-catechin supplementation leads to elicitation of disease resistance against Pseudomonas syringae DC3000 (hereafter DC3000). Intrigued by the unique hormesis response observed, we tested whether (+/-)-catechin indeed promotes growth responses in A. thaliana. In our hands, we observed no growth promotion responses of (+/-)-catechin against A. thaliana under in vitro or in soil conditions. We also evaluated the previously reported disease protecting properties of (+/-)-catechin in A. thaliana against DC3000. The systematic observations to evaluate disease protecting properties entailing colony counts, disease incidences and loss of chlorophyll studies showed no disease protecting properties of (+/-)-catechin. The transcriptional response for a marker pathogenesis related PR1 defense gene showed no induction post (+/-)-catechin supplementation. The cell death genes (ACD2 and CAD1) associated with programmed cell death revealed unchanged expression levels in plants treated with sub-MIC levels of (+/-)-catechin. Further, we report supplementation of sub-MIC levels of (+/-)-catechin negates any change in the expression of an auxin responsive gene. Our results refute the previous claims of growth and defense inducing effects of (+/-)-catechin, thus suggesting that a thorough reexamination is required to evaluate the hormetic effect of (+/-)-catechin under both controlled and natural conditions.
Sigma-Aldrich
(±)-Catechin hydrate
Pricing and availability is not currently available.