Merck
  • On the relevance of an in vitro reconstructed human epidermis model for drug screening in atopic dermatitis.
On the relevance of an in vitro reconstructed human epidermis model for drug screening in atopic dermatitis.
Experimental dermatology (2018-10-20)
Roland Hubaux, Coralie Bastin, Michel Salmon
ABSTRACT
Recent advances in the development of human-based in vitro models offer new tools for drug screening and mechanistic investigations of new therapeutic agents. However, there is a lack of evidence that disease models respond favourably to potential drug candidates. Atopic dermatitis (AD) is a very common disease associated with an altered skin barrier and chronic inflammation. Here, we demonstrate that the AD-like features of a reconstructed human epidermis (RHE) model treated with Th2 cytokines are reversed in the presence of molecules known to have a beneficial effect on damaged skin as a result of modulating various signalling cascades including the Liver X Receptors and JAK/STAT pathways. This work shows that standardized and reproducible RHE are relevant models for therapeutic research assessing new drug candidates aiming to restore epidermal integrity in an inflammatory environment.
Sigma-Aldrich
Anti-CA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.