Merck
In vitro antimicrobial activity of MSI-78, a magainin analog.
Antimicrobial agents and chemotherapy (1998-05-21)
P C Fuchs, A L Barry, S D Brown
ABSTRACT
MSI-78 is a cationic peptide with broad-spectrum antimicrobial activity and is being developed as a topical agent. We compared the in vitro activity of MSI-78 with those of ofloxacin and other antibiotics against fresh clinical isolates. Based on MIC distribution statistics, strains for which the MSI-78 MIC was < or = 64 micro/ml were assumed to be susceptible for purposes of this report. Of 411 aerobic isolates tested, 91% were susceptible to MSI-78, compared to 91% for ofloxacin and 92% for ciprofloxacin. Only enterococci consistently required > or = 64 microg of MSI-78/ml for inhibition. MSI-78 demonstrated bactericidal activity equivalent to that of ofloxacin. Of 61 anaerobes, 97% were susceptible to MSI-78. Of 10 isolates of Candida albicans, 3 were inhibited by MSI-78 at 24 h. Further studies of this compound appear to be warranted.
Sigma-Aldrich
Pexiganan acetate, ≥98% (HPLC)