Merck
  • Concise syntheses of the cruciferous phytoalexins brassilexin, sinalexin, wasalexins, and analogues: expanding the scope of the vilsmeier formylation.
Concise syntheses of the cruciferous phytoalexins brassilexin, sinalexin, wasalexins, and analogues: expanding the scope of the vilsmeier formylation.
The Journal of organic chemistry (2005-02-26)
M Soledade C Pedras, Mukund Jha
ABSTRACT
Efficient syntheses of the phytoalexins brassilexin, sinalexin, and analogues are demonstrated through the application of the Vilsmeier formylation to indoline-2-thiones followed by a new aqueous ammonia workup procedure. Similarly, a very concise two-pot synthesis of the phytoalexins wasalexins using sequential formylation-amination of indolin-2-ones is described. Remarkably, this novel aqueous ammonia workup allows the sequential one-pot formylation-amination, expanding substantially the scope of the Vilsmeier formylation of both indoline-2-thiones and indolin-2-ones. The examination of the formylation-amination reaction and optimization of conditions, as well as the syntheses and antifungal activities of several brassilexin analogues, are reported.
Sigma-Aldrich
1-Methylindoline-2-thione, ≥95%