Merck
  • Probing replacement of pyrophosphate via click chemistry; synthesis of UDP-sugar analogues as potential glycosyl transferase inhibitors.
Probing replacement of pyrophosphate via click chemistry; synthesis of UDP-sugar analogues as potential glycosyl transferase inhibitors.
Carbohydrate research (2009-02-24)
Kar Kheng Yeoh, Terry D Butters, Brendan L Wilkinson, Antony J Fairbanks
ABSTRACT
A series of potential UDP-sugar mimics were readily synthesised by copper(I) catalysed modified Huisgen cycloaddition of the corresponding alpha-propargyl glycosides with 5-azido uridine in aqueous solution. None of the compounds accessed displayed significant inhibitory activity at concentrations of up to 4.5mM in an assay against bovine milk beta-1,4-galactosyltransferase.