Merck
  • Experimental and theoretical studies on the functionalization reactions of 4-benzoyl-1,5-diphenyl-1H-pyrazole-3-carboxylic acid and acid chloride with 2,3-diaminopyridine.
Experimental and theoretical studies on the functionalization reactions of 4-benzoyl-1,5-diphenyl-1H-pyrazole-3-carboxylic acid and acid chloride with 2,3-diaminopyridine.
Molecules (Basel, Switzerland) (2007-11-17)
Ismail Yildirim, Fatma Kandemirli, Elif Demir
ABSTRACT
The 1H-pyrazole-3-carboxylic acid 2 was converted in good yield (69%) into the corresponding 1H-pyrazole-3-carboxamide 5 via reaction of the acid chloride 3 with 2,3- diaminopyridine (4). A different product, the 3H-imidazo[4,5-b] pyridine derivative 6, was formed from the reaction of 3 with 4 and base in benzene for 5 hours. The structures of the synthesized compounds were determined spectroscopically. The mechanism of the reaction between 3 and 4 was examined theoretically.
Sigma-Aldrich
2,3-Diaminopyridine, 95%