Merck
  • Thermal decomposition studies of the polyhedral oligomeric silsesquioxane, POSSh, and when it is impregnated with the metallocene bis(eta5-cyclopentadienyl)zirconium (IV) dichloride or immobilized on silica.
Thermal decomposition studies of the polyhedral oligomeric silsesquioxane, POSSh, and when it is impregnated with the metallocene bis(eta5-cyclopentadienyl)zirconium (IV) dichloride or immobilized on silica.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2008-01-29)
D Bianchini, I S Butler, M M Barsan, W Martens, R L Frost, G B Galland, J H Z dos Santos
ABSTRACT
Thermal decomposition studies of the free polyhedral oligomeric silsesquioxane, POSSh, and when this compound has been impregnated with Cp2ZrCl2 (Cp = eta5-C5H5) or immobilized on SiO2 were conducted using infrared emission spectroscopy (IES) over a 100-1000 degrees C temperature range and by thermogravimetric analysis (TGA). The organic groups in POSS(h) apparently decompose thermally into Si-CH3, Si-H and other fragments. Upon impregnation with Cp2ZrCl2, however, a different thermal decomposition pathway was followed and new infrared emission bands appeared in the 1000-900 cm(-1) region suggesting the formation of Si-O-Zr moieties. When immobilized on SiO2 and subjected to thermal decomposition, the POSSh compound lost its organic groups and the inorganic structure remaining was incorporated into the SiO2 framework.
Sigma-Aldrich
Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride solution, 42 wt. % in methanol