Merck
  • Annexin VI is a mannose-6-phosphate-independent endocytic receptor for bovine β-glucuronidase.
Annexin VI is a mannose-6-phosphate-independent endocytic receptor for bovine β-glucuronidase.
Experimental cell research (2011-06-16)
Alberto Ramírez-Mata, Colette Michalak, Guillermo Mendoza-Hernández, Alfonso León-Del-Río, Alfonso González-Noriega
ABSTRACT
Endocytosis and transport of bovine liver β-glucuronidase to lysosomes in human fibroblasts are mediated by two receptors: the well-characterized cation-independent mannose 6-phosphate receptor (IGF-II/Man6PR) and an IGF-II/Man6PR-independent receptor, which recognizes a Ser-Trp*-Ser sequence present on the ligand. The latter receptor was detergent extracted from bovine liver membranes and purified. LC/ESI-MS/MS analysis revealed that this endocytic receptor was annexin VI (AnxA6). Several approaches were used to confirm this finding. First, the binding of bovine β-glucuronidase to the purified receptor from bovine liver membranes and His-tagged recombinant human AnxA6 protein was confirmed using ligand-blotting assays. Second, western blot analysis using antibodies raised against IGF-II/Man6PR-independent receptor as well as commercial antibodies against AnxA6 confirmed that the receptor and AnxA6 were indeed the same protein. Third, double immunofluorescence experiments in human fibroblasts confirmed a complete colocalization of the bovine β-glucuronidase and the AnxA6 receptor on the plasma membrane. Lastly, two cell lines were stably transfected with a plasmid containing the cDNA for human AnxA6. In both transfected cell lines, an increase in cell surface AnxA6 and in mannose 6-phosphate-independent endocytosis of bovine β-glucuronidase was detected. These results indicate that AnxA6 is a novel receptor that mediates the endocytosis of the bovine β-glucuronidase.
Sigma-Aldrich
D-Mannose 6-phosphate disodium salt hydrate, ≥97.0% dry basis (enzymatic)
Sigma-Aldrich
D-Mannose 6-phosphate sodium salt, ≥98% (HPLC)