Merck
  • Identification of histidine residues involved in Zn(2+) binding to αA- and αB-crystallin by chemical modification and MALDI TOF mass spectrometry.
Identification of histidine residues involved in Zn(2+) binding to αA- and αB-crystallin by chemical modification and MALDI TOF mass spectrometry.
The protein journal (2012-08-15)
Srabani Karmakar, K P Das
ABSTRACT
α-Crystallin, a member of the small heat shock protein family is the major protein of mammalian eye lens and is a molecular chaperone. As there is no protein turn over in the lens, stability of α-crystallin is one of the most crucial factors for its survival and function. We previously reported that the molecular chaperone-like activity and stability of α-crystallin dramatically increased in the presence of Zn(2+) (Biochemistry, 2008). We also reported that each subunit of α-crystallin could bind multiple zinc ions through inter-subunit bridging giving rise to enhanced stability (Biopolymers, 2011). The amino acid residues involved in zinc binding were not known. Since cysteine residues were not responsible for binding to Zn(2+), we tried to identify the histidine residues bound to zinc ions. We modified recombinant αA- and αB-crystallin with diethylpyrocarbonate (DEPC) a histidine modifying reagent, in presence and absence of Zn(2+) followed by tryptic digestion. The residues modified by DEPC were identified through peptide mass matching by MALDI mass spectrometry. We have clearly identified H79, H107 and H115 of αA-crystallin and H104, H111 and H119 of αB-crystallin as the Zn(2+) binding residues. The significance of the histidine rich sequence region of α-crystallin for its stability is discussed.
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)