Merck
Metabolism of 1,3-dibromopropane.
Toxicology letters (1981-04-01)
S P James, M A Pue, D H Richards
ABSTRACT
Oral administration of 1,3-dibromopropane (2 mmol/kg) to rats resulted in a marked decrease in the level of hepatic glutathione (GSH). Sulphur-containing [14C]-metabolites were excreted in the bile of rats dose with 1,3-bromo[14C]propane and were subjected to enterohepatic cycling. After an oral dose of 1,3-dibromo[14C]propane, peak levels of radioactivity were rapidly attained in the blood and were maintained for several hours; approximately equal amounts of radioactive material were excreted in urine and expired air. Several radioactive metabolites were excreted in urine; a major metabolite was N-acetyl-S-[1-bromo-3-propyl]-cysteine.
Sigma-Aldrich
1,3-Dibromopropane, ReagentPlus®, 99%
Pricing and availability is not currently available.