Merck
  • Renal 21-hydroxylation of 19-hydroxy-progesterone to 19-hydroxy-deoxycorticosterone.
Renal 21-hydroxylation of 19-hydroxy-progesterone to 19-hydroxy-deoxycorticosterone.
Journal of steroid biochemistry (1989-11-01)
G T Griffing, M M Holbrook, F A Bencsath, J C Melby
ABSTRACT
19-Nor-deoxycorticosterone (19-nor-DOC) is a mineralocorticoid present in both rat and human urine, and it is elevated in some forms of experimental and human hypertension. Although the exact steps in the biosynthesis of 19-nor-DOC are uncertain, it is probably produced from a 19-oxygenated derivative of DOC, which undergoes 19-desmolation in the kidney. Since DOC biosynthesis is partly due to renal 21-hydroxylation of progesterone (Prog), we sought to determine whether a parallel pathway could exist for the biosynthesis of 19-hydroxy-DOC, a precursor to 19-nor-DOC. In order to test this hypothesis, authentic 19-hydroxy-progesterone was incubated with homogenized renal tissues from either rat or human sources. Formation of 19-hydroxy-DOC was found to be the major metabolite in both rat and human incubations, as demonstrated by an HPLC retention time identical to authentic 19-hydroxy-DOC. 19-Hydroxy-DOC formation was further verified by GC/MS analysis with highly sensitive selected ion recording. Since it has been demonstrated that the placenta can convert progesterone to 19-hydroxy-progesterone, the renal 21-hydroxylation of 19-hydroxy-progesterone to 19-hydroxy-DOC could be an alternate pathway of 19-nor-DOC production especially during pregnancy.