Merck
2,7-Dimethoxynaphthalene.
Acta crystallographica. Section C, Crystal structure communications (1989-08-15)
P Prince, F R Fronczek, R D Gandour
ABSTRACT
C12H12O2, Mr = 188.2, orthorhombic, P2(1)2(1)2(1), a = 6.109 (3), b = 8.235 (2), c = 19.713 (3) A, V = 991.8 (9) A3, Z = 4, Dx = 1.260 g cm-3, lambda(Mo K alpha) = 0.71073 A, mu = 0.79 cm-1, F(000) = 400, T = 293 K, R = 0.047 for 737 observations (of 1337 unique data). The average deviation from planarity is 0.013 (3) A with a maximum of 0.027 (3) A for the fused rings. Steric interaction between the methyl groups and the adjacent H atoms is responsible for opening the angles between the methoxyl groups and the rings. The CH3O-C-C angles are 125.5 (3) degrees [125.0 (3) degrees] where the methyl group is syn to the hydrogen, and 114.3 (3) degrees [114.2 (3) degrees] where it is anti.
Sigma-Aldrich
2,7-Dimethoxynaphthalene, 98%
Pricing and availability is not currently available.