Merck
  • Determination of immunoglobulin G in bovine colostrum and milk by direct biosensor SPR-immunoassay.
Determination of immunoglobulin G in bovine colostrum and milk by direct biosensor SPR-immunoassay.
Journal of AOAC International (2003-05-02)
Harvey E Indyk, Enrico L Filonzi
ABSTRACT
An automated biosensor surface-plasmon resonance-based assay was developed for the determination of immunoglobulin G (IgG) in bovine milk and colostrum with either goat or rabbit antibovine IgG or protein G used as detecting molecule. The method is configured as a direct and nonlabeled immunoassay, with quantitation against an authentic IgG calibrant. Whole colostrum or milk is prepared for analysis by dilution into buffer. Analysis conditions, including ligand immobilization, flowrate, contact time, and regeneration, were optimized, and nonspecific binding was evaluated. Performance parameters included working range of 15-10 000 ng/mL, method detection limit of 0.08 mg/mL, overall instrument response reproducibility relative standard deviation (RSD(R)) of 0.47%, mean between-run RSD(R) of 10.5% for colostrum, and surface stability over 200 analyses. The proposed method was compared with independent alternative methods. The technique was applied to the measurement of IgG content during early lactation transition from colostrum to milk, as well as in consumer milk, colostrum, and hyperimmune milk powders.
Sigma-Aldrich
γ-Globulins from bovine blood, ≥99% (agarose gel electrophoresis)
Pricing and availability is not currently available.