- Cell-cycle dependent mutation of human lymphoblasts: bromodeoxyuridine and butyl methanesulfonate.
Cell-cycle dependent mutation of human lymphoblasts: bromodeoxyuridine and butyl methanesulfonate.
Mutation research (1991-09-11)
H Hoppe, K M Call, P M Leong, W G Thilly
PMID1658642
ABSTRACT
Cells of the human lymphoblast line WI-L2 and its derivative TK-6 were synchronized by centrifugal elutriation and cell-cycle dependent mutation to 6TGR (HPRT) and OUAR (Na+, K+ ATPase) measured. Bromodeoxyuridine induced 6TGR and OUAR mutations within S phase while butylmethyl-sulfonate induced mutation displayed no cell-cycle dependence. The data indicate that centrifugal elutriation is a facile means to obtain a useful degree of synchrony for these cell lines.
Product Description
Butyl methanesulfonate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.