Merck
  • Vildagliptin added to sulfonylurea improves glycemic control without hypoglycemia and weight gain in Chinese patients with type 2 diabetes mellitus.
Vildagliptin added to sulfonylurea improves glycemic control without hypoglycemia and weight gain in Chinese patients with type 2 diabetes mellitus.
Journal of diabetes (2014-05-16)
Wenying Yang, Xiaoping Xing, Xiaofeng Lv, Yiming Li, Jianhua Ma, Guoyue Yuan, Feifei Sun, Wei Wang, Michael Woloschak, Valentina Lukashevich, Plamen Kozlovski, Wolfgang Kothny
ABSTRACT
The aim of the present study was to assess the efficacy and safety of vildagliptin as add-on to sulfonylurea therapy in Chinese patients with type 2 diabetes mellitus (T2DM) inadequately controlled on sulfonylurea monotherapy. The 24-week randomized double-blind placebo-controlled study compared vildagliptin 50 mg, q.d., with placebo as add-on to glimepiride in T2DM patients who were inadequately controlled (HbA1c 7.5%-11.0% [58-97 mmol/mol]) on a stable dose of sulfonylurea for ≥12 weeks before study entry. In all, 279 patients were randomized to receive either vildagliptin (n = 143) or placebo (n = 136). At baseline, overall mean age was 58.5 years, body weight 68.1 kg, duration of diabetes 6.9 years and daily glimepiride dose 3.3 mg. After 24 weeks, the adjusted mean change (AMΔ) in HbA1c was -0.7% (-8 mmol/mol; baseline 8.6%, 70 mmol/mol) in the vildagliptin group and -0.2% (-2 mmol/mol; baseline 8.7%, 72 mmol/mol) in the placebo group, with a treatment difference of -0.5% (-5 mmol/mol; P < 0.001). The between-group difference in AMΔ in fasting plasma glucose was -0.4 mmol/L (P = 0.160). There was a slight, but not significant, decrease in body weight in both groups. No hypoglycemic events were reported in either group, including those patients reaching HbA1c <7.0%. Patients in the vildagliptin and placebo groups reported low and comparable incidences of adverse events (14.0% vs. 17.8%) and serious adverse events (0.7% in each group). Vildagliptin 50 mg, q.d., added to sulfonylurea monotherapy is effective in Chinese patients with T2DM, without increasing the risk of hypoglycemia and weight gain.
Product Description
USP
Glimepiride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glimepiride, ≥98% (HPLC), solid
Glimepiride, European Pharmacopoeia (EP) Reference Standard