- Highly efficient synthesis of cyclic ureas from CO2 and diamines by a pure CeO2 catalyst using a 2-propanol solvent.
Highly efficient synthesis of cyclic ureas from CO<SUB>2</SUB> and diamines by a pure CeO<SUB>2</SUB> catalyst using a 2-propanol solvent.
Green Chemistry (2013)
Tamura M, et al.