Merck
  • Biocompatible molecularly imprinted polymers for the voltage regulated uptake and release of L-glutamate in neutral pH solutions.
Biocompatible molecularly imprinted polymers for the voltage regulated uptake and release of L-glutamate in neutral pH solutions.
Biosensors & bioelectronics (2010-08-07)
E von Hauff, K Fuchs, D Ch Hellmann, J Parisi, R Weiler, C Burkhardt, U Kraushaar, E Guenther
ABSTRACT
A biocompatible device for the voltage dependent uptake and release of the neural transmitter L-glutamate in neutral pH solutions is demonstrated. The device consists of a gold electrode coated with molecularly imprinted, overoxidised polypyrrole (oPPy). It is shown here that oPPy can behave as an anion exchanger in neutral pH. The voltage dependent uptake and release of glutamate from the oPPy as well as the enantioselectivity of the polymer layer for L-glutamate over D-glutamate are investigated in neutral pH solutions using electrochemical quartz crystal microbalance techniques. The biocompatibility of the oPPy layer is demonstrated using retinae from young rats. The retinae were isolated and the dissociated cells were kept in culture for up to 1-week. The cells were exposed to the oPPy layers for 3 days, and there is no significant difference in the survival rate between the cells cultured on the oPPy layers and the control samples. Additionally the cell-polymer interface from cells grown directly on the oPPy layers is investigated using electron microscopy.
Product Description
Supelco
L-Glutamic acid monosodium salt hydrate, analytical standard
Pricing and availability is not currently available.