Merck
  • 184: Toxic effects of antibiotics routinely used in cell culture on primary cultures of dissociated mouse brain cells.
184: Toxic effects of antibiotics routinely used in cell culture on primary cultures of dissociated mouse brain cells.
Pediatric Research (1976)
Ammon F