Merck
  • A convenient and adjustable surface-modified complex containing poly-L-glutamic acid conjugates as a vector for gene delivery.
A convenient and adjustable surface-modified complex containing poly-L-glutamic acid conjugates as a vector for gene delivery.
International journal of nanomedicine (2008-08-09)
Ye Sun, Yuzhao Tang, Maoquan Chu, Shuxian Song, Yanfei Xin
ABSTRACT
In order to quantify the amount of ligands or poly(ethylene glycol) (PEG) on each vector, here we developed a system in which poly-L-glutamic acid (PLG) was used as surface modification loading backbone, to which one PEG (MW 5000, 10000, 20000) or epidermal growth factor (EGF) was linked. The PLG conjugates can electro-statically adsorb upon DNA/ polycation complex with positive charge, and, the amount of EGF or PEG on the surface of complexes could be varied. We have made a series of complexes containing the various PLG conjugates and examined their physicochemical properties, and made a comparison of properties and transfection efficiency between these complexes. EGF- and PEG-modified complexes showed 10-25-folds higher cell transfection efficiency than unmodified complexes in medium with or without serum.
Sigma-Aldrich
4-(N-Maleimidomethyl)cyclohexanecarboxylic acid N-hydroxysuccinimide ester, ≥98%, powder