Merck
  • Systemic and spinal administration of FAAH, MAGL inhibitors and dual FAAH/MAGL inhibitors produce antipruritic effect in mice.
Systemic and spinal administration of FAAH, MAGL inhibitors and dual FAAH/MAGL inhibitors produce antipruritic effect in mice.
Archives of dermatological research (2016-04-30)
Ozgur Yesilyurt, Mutlu Cayirli, Yusuf Serdar Sakin, Melik Seyrek, Ahmet Akar, Ahmet Dogrul
ABSTRACT
The increase of endocannabinoid tonus by inhibiting fatty acid amide hydrolase (FAAH) or monoacylglycerol lipase (MAGL) represents a promising therapeutic approach in a variety of disease to overcome serious central side effects of exocannabinoids. Recent studies reported that systemic administration of FAAH and MAGL inhibitors produce antipruritic action. Dual FAAH/MAGL inhibitors have also been described to get enhanced endocannabinoid therapeutic effect. In this study, we examined and compared dose-related antipruritic effects of systemic (intraperitoneal; ip) or intrathecal (it) administration of selective FAAH inhibitor PF-3845 (5, 10, and 20 mg/kg, i.p.; 1, 5, and 10 µg, i.t.), MAGL inhibitor JZL184 (4, 20, and 40 mg/kg, i.p.; 1, 5, and 10 µg, i.t.) and dual FAAH/MAGL inhibitor JZL195 (2, 5, and 20 mg/kg, i.p.; 1, 5, and 10 µg, i.t.) on serotonin (5-HT)-induced scratching model. Serotonin (25 μg) was injected intradermally in a volume of 50 μl into the rostral part of skin on the back of male Balb-C mice. Both systemic or intrathecal administration of PF-3845, JZL184 or JZL195 produced similar dose-dependent antipruritic effects. Our results suggest that endocannabinoid-degrading enzymes FAAH and MAGL are involved in pruritic process at spinal level. FAAH, MAGL or dual FAAH/MAGL inhibitors have promising antipruritic effects, at least, in part through spinal site of action.
Sigma-Aldrich
JZL195, ≥98% (HPLC)
Pricing and availability is not currently available.