Merck
  • Mutations in the mitochondrial GTPase mitofusin 2 cause Charcot-Marie-Tooth neuropathy type 2A.
Mutations in the mitochondrial GTPase mitofusin 2 cause Charcot-Marie-Tooth neuropathy type 2A.
Nature genetics (2004-04-06)
Stephan Züchner, Irina V Mersiyanova, Maria Muglia, Nisrine Bissar-Tadmouri, Julie Rochelle, Elena L Dadali, Mario Zappia, Eva Nelis, Alessandra Patitucci, Jan Senderek, Yesim Parman, Oleg Evgrafov, Peter De Jonghe, Yuji Takahashi, Shoij Tsuji, Margaret A Pericak-Vance, Aldo Quattrone, Esra Battaloglu, Alexander V Polyakov, Vincent Timmerman, J Michael Schröder, Jeffery M Vance, Esra Battologlu