- Synthesis, fluorescence-sensing and molecular logic of two water-soluble 1,8-naphthalimides.
Synthesis, fluorescence-sensing and molecular logic of two water-soluble 1,8-naphthalimides.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-04-23)
Nikolai I Georgiev, Margarita D Dimitrova, Anelia Ts Mavrova, Vladimir B Bojinov
PMID28432919
ABSTRACT
Two novel highly water-soluble fluorescence sensing 1,8-naphthalimides are synthesized and investigated. The novel compounds are designed on the "fluorophore-receptor