Merck
  • Hepatitis B virus X protein induces hepatic stem cell-like features in hepatocellular carcinoma by activating KDM5B.
Hepatitis B virus X protein induces hepatic stem cell-like features in hepatocellular carcinoma by activating KDM5B.
World journal of gastroenterology (2017-06-02)
Xuyang Wang, Naoki Oishi, Tetsuro Shimakami, Taro Yamashita, Masao Honda, Seishi Murakami, Shuichi Kaneko
ABSTRACT
To determine the role of hepatitis B virus X protein (HBx), HBx in regulating hepatic progenitor cell (HPC)-like features in hepatocellular carcinoma (HCC) and the underlying molecular mechanisms. We used a retrovirus vector to introduce wild type HBx or empty vector into HepG2 cells. We then used these cells to analyze cell proliferation, senescence, transformation, and stem-like features. Gene expression profiling was carried out on Affymetrix GeneChip Human U133A2.0 ver.2 arrays according to the manufacturer's protocol. Unsupervised hierarchical clustering analysis and Class Comparison analysis were performed by BRB-Array Tools software Version 4.2.2. A total of 238 hepatitis B virus (HBV)-related HCC patients' array data were used for analyzing clinical features. The histone demethylase KDM5B was significantly highly expressed in HBV-related HCC cases ( HBx activates the histone demethylase KDM5B and induces HPC-like features in HCC. Histone demethylases KDM5B may be an important therapeutic target against HBV-related HCC cases.
Sigma-Aldrich
MISSION® esiRNA, targeting human MYC
Pricing and availability is not currently available.