Merck
  • Highly N-doped microporous carbon nanospheres with high energy storage and conversion efficiency.
Highly N-doped microporous carbon nanospheres with high energy storage and conversion efficiency.
Scientific reports (2017-11-02)
Cheolho Kim, Kiwon Kim, Jun Hyuk Moon
ABSTRACT
Porous carbon spheres (CSs) have distinct advantages in energy storage and conversion applications. We report the preparation of highly monodisperse N-doped microporous CSs through the carbonization of polystyrene-based polymer spheres and subsequent activation. The N-doped microporous CSs have a remarkably high N-doping content, over 10%, and high BET surface area of 884.9 m
Sigma-Aldrich
Valeronitrile, 99.5%
Pricing and availability is not currently available.