Merck
Prostaglandin D
Scientific reports (2017-12-17)
Shingo Maeda, Tatsuro Nakamura, Hiroaki Harada, Yuri Tachibana, Kosuke Aritake, Tatsuo Shimosawa, Yutaka Yatomi, Takahisa Murata
ABSTRACT
Food allergy is immediate hypersensitive reactions to ingested foods. Since early diagnosis is effective for disease control, development of an objective diagnostic index is required. Using mediator-lipidomics, we found that levels of the urinary prostaglandin D