Merck
Identification of novel PDEδ interacting proteins.
Bioorganic & medicinal chemistry (2017-09-25)
Philipp Küchler, Gunther Zimmermann, Michael Winzker, Petra Janning, Herbert Waldmann, Slava Ziegler
ABSTRACT
Prenylation is a post-translational modification that increases the affinity of proteins for membranes and mediates protein-protein interactions. The retinal rod rhodopsin-sensitive cGMP 3',5'-cyclic phosphodiesterase subunit delta (PDEδ) is a prenyl binding protein that is essential for the shuttling of small GTPases between different membrane compartments and, thus, for their proper functioning. Although the prenylome comprises up to 2% of the mammalian proteome, only few prenylated proteins are known to interact with PDEδ. A proteome-wide approach was employed to map the PDEδ interactome among the prenylome and revealed RAB23, CDC42 and CNP as novel PDEδ interacting proteins. Moreover, PDEδ associates with the lamin A mutant progerin in a prenyl-dependent manner. These findings shed new light on the role of PDEδ in binding (and regulating) prenylated proteins in cells.
Sigma-Aldrich
Deltarasin trihydrochloride, ≥98% (HPLC)
Pricing and availability is not currently available.