Merck
  • Tissue expression and sero-reactivity of tumor-specific antigens in colorectal cancer.
Tissue expression and sero-reactivity of tumor-specific antigens in colorectal cancer.
Cancer letters (2004-05-15)
Axel Gerhardt, Dirk Usener, Michael Keese, Jörg Sturm, Dirk Schadendorf, Stefan Eichmüller
ABSTRACT
The expression of 14 individual and two groups of tumor antigens was characterized for colorectal carcinoma by RT-PCR using 26 colorectal carcinoma specimens, eight cell lines, six samples of patients with inflammatory bowl diseases, and nine specimens from different locations of an individual patient with a metastasized rectal carcinoma. The most frequently detected mRNAs were MAGE-A1 (58%), GAGE-3-7 (54%), and cTAGE-5a (31%). At medium frequencies (12-19%) we found cTAGE-1, MAGE-A2, se57-1, RAGE-4, and GAGE-1,2,8, while other tumor antigens were expressed rarely (<9%). 85% of the samples were positive for at least one of the most frequently expressed antigens. Using a secondary SEREX approach and sera of eight colorectal cancer patients we found reactive antibodies against recombinant cTAGE-1 (2 sera), se57-1 (2), truncated GAGE (1), and MAGE-A1 (1). We conclude that certain cancer-germline genes can be detected in colorectal cancer and might therefore be promising targets for immunotherapy.