All Photos(1)

TMRES-216ML

Sigma-Aldrich

TurboMix Resolving Solution, 216 mL

sterility

0.2 μm filtered

form

liquid

feed ratio

29:1

storage condition

protect from light

storage temp.

2-8°C

General description

TurboMix Resolving Solution is a 20% acrylamide buffer solution used to make bis-tris polyacrylamide gels. It can be diluted with DI water to make any gel percentage between 8% and 15%. After dilution, add APS and TEMED and then pour gels to the desired height.

Application

Premixed resolving gel solution for casting bis-tris polyacrylamide gels.

Legal Information

TURBOMIX is a trademark of Merck KGaA

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificate of Analysis

Certificate of Origin

Articles

Bis Tris Polyacrylamide Gel Electrophoresis Technology

Bis-Tris gels and buffers for superior protein resolution compared to traditional tris-glycine gels.

Protocols

Hand-casting gels for PAGE and SDS-PAGE using TurboMix™ Bis-Tris Gel Casting Kits

Overview of polyacrylamide gel chemistry and detailed instructions for hand-casting polyacrylamide gels using TurboMix™ Bis-Tris Gel Casting Kits

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service